missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylbenzenesulfonyl chloride, 97%

Product Code. 15425687
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15425687 1 g 1g
15435687 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425687 Supplier Thermo Scientific Alfa Aesar Supplier No. H32912.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 34981-38-9 | C7H6Cl2O2S | 225.083 g/mol

5-Chloro-2-methylbenzenesulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-2-methylbenzenesulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34981-38-9
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.083
MDL Number MFCD09742738
InChI Key VHBFBNCERXCECQ-UHFFFAOYSA-N
Synonym 4-chlorotoluene-2-sulphonyl chloride, 4-chlorotoluene-2-sulphonylchloride, 2-chlorosulfonyl-4-chlorotoluene, 5-chloro-2-methylbenzene-1-sulfonyl chloride, 5-chloro-2-methylbenzenesulfonylchloride, benzenesulfonyl chloride, 5-chloro-2-methyl, 5-chloro-2-methylbenzensulfonyl chloride, 5-chloro-2-methylphenylsulfonyl chloride, 3-chloro-6-methyl-benzenesulfonyl chloride, benzenesulfonylchloride, 5-chloro-2-methyl
PubChem CID 100415
IUPAC Name 5-chloro-2-methylbenzenesulfonyl chloride
SMILES CC1=C(C=C(C=C1)Cl)S(=O)(=O)Cl

Specifications

Refractive Index 1.5697
Quantity 1 g
UN Number UN3265
Solubility Information Slightly soluble in water.
Formula Weight 225.09
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-methylbenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.