missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-nitroaniline, 97%

Product Code. 11403584
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403584 25 g 25g
11493574 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11403584 Supplier Thermo Scientific Alfa Aesar Supplier No. A11241.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1635-61-6 | C6H5ClN2O2 | 172.568 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1635-61-6
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD00007776
InChI Key ZCWXYZBQDNFULS-UHFFFAOYSA-N
Synonym 2-nitro-5-chloroaniline, benzenamine, 5-chloro-2-nitro, 3-chloro-6-nitroaniline, 5-chloro-2-nitrobenzeneamine, aniline, 5-chloro-2-nitro, 2-amino-4-chloro-nitrobenzene, unii-656xjm4con, 5-chloro-2-nitrobenzenamine, ccris 8934, 5-chloro-2-nitro aniline
PubChem CID 74218
IUPAC Name 5-chloro-2-nitroaniline
SMILES C1=CC(=C(C=C1Cl)N)[N+](=O)[O-]

Specifications

Melting Point 124°C to 128°C
Odor Weak, characteristic
Quantity 25 g
UN Number UN2237
Beilstein 2210201
Formula Weight 172.57
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-nitroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.