missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-nitroaniline, TRC

Product Code. 30350133
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350133 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350133 Supplier Toronto Research Chemicals Supplier No. TRCC3737705G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chloro-2-nitroaniline
CAS 1635-61-6
Molecular Formula C6H5ClN2O2
Quantity 5 g
Synonym 5-Chloro-2-nitro-benzenamine, 5-Chloro-2-nitro-aniline, 1-Nitro-2-amino-4-chlorobenzene, 2-Amino-4-chloro-1-nitrobenzene, 2-Amino-4-chloronitrobenzene, 2-Nitro-5-chloroaniline, 3-Amino-4-nitrochlorobenzene, 3-Chloro-6-nitroaniline, 5-Chlor-2-nitroanilin, 5-Chloro-2-nitroaniline, 5-Chloro-2-nitrobenzenamine, 5-Chloro-2-nitrophenylamine, NSC 400866
InChI Formula InChI=1S/C6H5ClN2O2/c7-4-1-2-6(9(10)11)5(8)3-4/h1-3H,8H2
SMILES Nc1cc(Cl)ccc1[N+](=O)[O-]
IUPAC Name 5-chloro-2-nitroaniline
Molecular Weight (g/mol) 172.57
Formula Weight 172.004
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.