missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-pentanone ethylene ketal, 97%

Product Code. 10184320 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10184320 25 g Glass bottle 25g
10563911 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10184320 Supplier Thermo Scientific Acros Supplier No. 166040250
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5978-08-5 | C7H13ClO2 | 164.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5978-08-5
Molecular Formula C7H13ClO2
Molecular Weight (g/mol) 164.63
MDL Number MFCD00003217
InChI Key OFERIRWCHSOJJT-UHFFFAOYSA-N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 73°C to 76°C (7.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4490 to 1.451
Quantity 25 g
Specific Gravity 1.09
Synonym 6-hydroxymethyl oxane-2,3,4,5-tetrol hydrate, d-galactose hydrate, dextrose monohydrate, usp, .alpha.-d-glucose hydrate, dextrose, u.s.p.
SMILES CC1(CCCCl)OCCO1
IUPAC Name (2R,3S,4R,5R,6S)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol;hydrate
PubChem CID 133126654
Formula Weight 164.63
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 5-Chloro-2-pentanone ethylene ketal, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.