missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-pentanone ethylene ketal, 97%

Product Code. 10563911 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10563911 5 g Glass bottle 5g
10184320 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10563911 Fornitore Thermo Scientific Acros N. del fornitore 166040050
 Restricted Item
Estimated Shipment: 09-12-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 5978-08-5 | C7H13ClO2 | 164.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 5978-08-5
Molecular Formula C7H13ClO2
Molecular Weight (g/mol) 164.63
MDL Number MFCD00003217
InChI Key OFERIRWCHSOJJT-UHFFFAOYSA-N

Specifica

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 73°C to 76°C (7.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4490 to 1.451
Quantity 5 g
Specific Gravity 1.09
Synonym 2-3-chloropropyl-2-methyl-1,3-dioxolane, 5-chloro-2-pentanone ethylene ketal, 2,2-ethylenedioxy-5-chloropentane, 1,3-dioxolane, 2-3-chloropropyl-2-methyl, 5-chloro-2-pentanoneethyleneketal, acmc-20am4f, oferirwchsojjt-uhfffaoysa, 1-chloro-4-pentanone ethylene ketal, 1, 2-3-chloropropyl-2-methyl, 5-chloro-2-pentanone ethylene acetal
SMILES CC1(OCCO1)CCCCl
IUPAC Name 2-(3-chloropropyl)-2-methyl-1,3-dioxolane
PubChem CID 80079
Formula Weight 164.63
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 5-Chloro-2-pentanone ethylene ketal, 97%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.