missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-Thiophenecarboxylic Acid 4-Nitrophenyl Ester, TRC

Product Code. 30356453
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30356453 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30356453 Supplier Toronto Research Chemicals Supplier No. TRCC424380250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chloro-2-Thiophenecarboxylic Acid 4-Nitrophenyl Ester
CAS 1450877-56-1
Molecular Formula C11H6ClNO4S
Quantity 250 mg
Synonym 2-Thiophenecarboxylic acid, 5-chloro-, 4-nitrophenyl ester (ACI), 4-Nitrophenyl 5-chloro-2-thiophenecarboxylate (ACI), 4-Nitrophenyl 5-chlorothiophene-2-carboxylate
InChI Formula InChI=1S/C11H6ClNO4S/c12-10-6-5-9(18-10)11(14)17-8-3-1-7(2-4-8)13(15)16/h1-6H
SMILES O[13C]([13C@@]([15NH2])([H])[13C]([H])([H])[13C]([H])([H])[15N]=[N]=N)=O.Cl
IUPAC Name (4-nitrophenyl) 5-chlorothiophene-2-carboxylate
Molecular Weight (g/mol) 283.69
Formula Weight 282.9706
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.