missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-2-toluidine, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Chloro-2-methylaniline |
| CAS | 95-79-4 |
| Molecular Formula | C7 H8 Cl N |
| Quantity | 10 g |
| Synonym | Fast Red KB Base (6CI), o-Toluidine, 5-chloro- (8CI), 5-Chloro-2-methylbenzenamine, 2-Amino-4-chlorotoluene, 2-Methyl-5-chloroaniline, 3-Chloro-6-methylaniline, 4-Chloro-2-aminotoluene, 5-Chloro-2-methylaniline, 5-Chloro-2-methylphenylamine, 5-Chloro-2-toluidine, 5-Chloro-o-toluidine, Acco Fast Red KB base, Ansibase Red KB, Azoene Fast Red KB base, Fast Red KB amine, Fast Red KB salt, Fast Red KB salt Supra, Fast Red KB-T Base, Fast Red KBS salt, Genazo Red KB soln, Hiltonil Fast Red KB base, Metrogen Red Former KB soln, NSC 7094, Naphthosol Fast Red KB base, Pharmazoid Red KB, Red KB base, Spectrolene Red KB, Stable Red KB base |
| InChI Formula | InChI=1S/C7H8ClN/c1-5-2-3-6(8)4-7(5)9/h2-4H,9H2,1H3 |
| SMILES | Cc1ccc(Cl)cc1N |
| IUPAC Name | 5-chloro-2-methylaniline |
| Molecular Weight (g/mol) | 141.6 |
| Formula Weight | 141.0345 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
¿Detecta una oportunidad de mejora?