missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-3-(chloromethyl)-1,2,4-thiadiazole, 97%, Thermo Scientific™

Product Code. 10556524
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10556524 5 g Amber Glass Bottle 5g
10161032 250 mg Amber Glass Bottle 250mg
10009531 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10556524 Supplier Thermo Scientific Maybridge Supplier No. SEW03081DE
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 74461-64-6
Molecular Formula C3H2Cl2N2S
Molecular Weight (g/mol) 169.02
MDL Number MFCD00096470
InChI Key FFFMIFUBJZBTCG-UHFFFAOYSA-N
Synonym 5-chloro-3-chloromethyl-1,2,4-thiadiazole, 5-chloro-3-chloromethyl-1,2,4 thiadiazole, 3-chloromethyl-5-chloro-1,2,4 thiadiazole, 3-chloromethyl-5-chloro-1,2,4-thiadiazole, 1,2,4-thiadiazole,5-chloro-3-chloromethyl, 1,2,4-thiadiazole, 5-chloro-3-chloromethyl-7ci, 9ci
PubChem CID 332610
IUPAC Name 5-chloro-3-(chloromethyl)-1,2,4-thiadiazole
SMILES ClCC1=NSC(Cl)=N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 169.03
Percent Purity 97%
Chemical Name or Material 5-chloro-3-(chloromethyl)-1, 2, 4-thiadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.