missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-3-phenyl-2,1-benzisoxazole, 97%, Thermo Scientific™

Product Code. 10381800 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10381800

Brand: Thermo Scientific Acros 438380050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 719-64-2
Molecular Formula C13H8ClNO
Molecular Weight (g/mol) 229.67
InChI Key MUHJZJKVEQASGY-UHFFFAOYSA-N
Synonym 5-chloro-3-phenylbenzo c isoxazole, 5-chloro-3-phenylanthranil, 5-chloro-3-phenyl-2,1-benzisoxazole, 3-phenyl-5-chloroanthranil, pubchem8713, acmc-209okx, maybridge1_004397, 2, 5-chloro-3-phenyl, 5-chloro-3-phenyl-benzo c isoxazole, 3-phenyl-5-chloro-2,1-benzisoxazole
PubChem CID 347291
IUPAC Name 5-chloro-3-phenyl-2,1-benzoxazole
SMILES C1=CC=C(C=C1)C2=C3C=C(C=CC3=NO2)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 115.3°C to 116.1°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 5 g
Formula Weight 229.67
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-3-phenyl-2, 1-benzisoxazole
Show More Show Less
compliance-icons
Product Identifier
  • 5-Chloro-3-phenyl-2,1-benzisoxazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.