missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-4-fluoro-2-nitroaniline, 98%, Thermo Scientific™

Product Code. 10272802 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10272802 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10272802 Supplier Thermo Scientific Acros Supplier No. 355490010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 104222-34-6
CAS Min % 97.5
CAS Max % 100.0
Melting Point 142°C to 145°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H4ClFN2O2
MDL Number MFCD00052698
Quantity 1 g
Synonym 2-nitro-4-fluoro-5-chloroaniline, benzenamine, 5-chloro-4-fluoro-2-nitro, benzenamine,5-chloro-4-fluoro-2-nitro, 5-chloro-4-fluoro-2-nitrophenylamine, pubchem8452, acmc-2098bg, timtec-bb sbb003647, buttpark 45\03-11, 3-chloro-4-fluoro-6-nitroaniline, 5-chloro-4-fluoro-2-nitro-aniline
InChI Key VRJKEIWZSOHDOH-UHFFFAOYSA-N
SMILES C1=C(C(=CC(=C1Cl)F)[N+](=O)[O-])N
IUPAC Name 5-chloro-4-fluoro-2-nitroaniline
Molecular Weight (g/mol) 190.56
PubChem CID 517835
Formula Weight 190.56
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-4-fluoro-2-nitroaniline
Show More Show Less
compliance-icons
Product Identifier
  • 5-Chloro-4-fluoro-2-nitroaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.