missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-4-nitrothiophene-2-sulfonyl chloride, 97%, Maybridge

Product Code. 10523042
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10523042 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10523042 Supplier Thermo Scientific Maybridge Supplier No. KM02596DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 58457-24-2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C4HCl2NO4S2
MDL Number MFCD00067978
Quantity 1 g
Synonym 2-chloro-3-nitrothiophene-5-sulphonyl chloride, 2-chloro-3-nitrothiophene-5-sulfonyl chloride, 5-chloro-4-nitro-thiophene-2-sulfonyl chloride, chloro 5-chloro-4-nitro 2-thienyl sulfone, pubchem5498, 5-chloro-4-nitrothiophene-2-sulfonylchloride, acmc-20am48, 2-chlorosulphonyl-4-nitro-5-chlorothiophene, 5-chloro-4-nitro-2-thienylsulfonyl chloride
InChI Key SGHAHFHTASPNKN-UHFFFAOYSA-N
SMILES C1=C(SC(=C1[N+](=O)[O-])Cl)S(=O)(=O)Cl
IUPAC Name 5-chloro-4-nitrothiophene-2-sulfonyl chloride
Molecular Weight (g/mol) 262.075
PubChem CID 2735783
Formula Weight 262.09
Percent Purity 97%
Chemical Name or Material 5-Chloro-4-nitrothiophene-2-sulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.