missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-8-hydroxyquinoline, 95%

Product Code. 11499887
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499887 50 g 50g
11409897 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499887 Supplier Thermo Scientific Alfa Aesar Supplier No. B22856.18
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 130-16-5 | C9H6ClNO | 179.603 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130-16-5
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.603
MDL Number MFCD00006788
InChI Key CTQMJYWDVABFRZ-UHFFFAOYSA-N
Synonym 5-chloro-8-hydroxyquinoline, cloxiquine, cloxyquin, 5-chloro-8-quinolinol, dermofongin, 5-chlorooxine, 8-quinolinol, 5-chloro, chlorisept, dermofungin, 5-chloro-8-oxyquinoline
PubChem CID 2817
IUPAC Name 5-chloroquinolin-8-ol
SMILES C1=CC2=C(C=CC(=C2N=C1)O)Cl

Specifications

Melting Point 122°C to 124°C
Quantity 50 g
Beilstein 5289
Sensitivity Light sensitive
Merck Index 14,2422
Formula Weight 179.61
Percent Purity 95%
Chemical Name or Material 5-Chloro-8-hydroxyquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.