missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorobenzo-2,1,3-thiadiazole, 98%

Product Code. 11493827
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493827 1 g 1g
11403837 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493827 Supplier Thermo Scientific Alfa Aesar Supplier No. A18286.03
 Restricted Item
Estimated Shipment: 30-12-2026
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 2207-32-1 | C6H3ClN2S | 170.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 2207-32-1
Molecular Formula C6H3ClN2S
Molecular Weight (g/mol) 170.61
MDL Number MFCD00174269
InChI Key VRNJWKISMWDTAY-UHFFFAOYSA-N
Synonym 5-chlorobenzo c 1,2,5 thiadiazole, 5-chlorobenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole, 5-chloro, 5-chlorobenzo c 1,2,5-thiadiazole, acmc-1cipd, maybridge1_004942, 5-chloranyl-2,1,3-benzothiadiazole, 3-chloro-8-thia-7,9-diazabicyclo 4.3.0 nona-2,4,6,9-tetraene
PubChem CID 284123
IUPAC Name 5-chloro-2,1,3-benzothiadiazole
SMILES ClC1=CC2=NSN=C2C=C1

Specifikationer

Melting Point 55°C to 57°C
Quantity 1 g
Formula Weight 170.62
Percent Purity 98%
Chemical Name or Material 5-Chlorobenzo-2,1,3-thiadiazole

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.