missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorobenzo[b]thiophene-3-acetic acid, 97%

Product Code. 11333396
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333396 250 mg 250mg
11323396 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333396 Supplier Thermo Scientific Alfa Aesar Supplier No. L12836.MD
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 17266-30-7 | C10H7ClO2S | 226.674 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17266-30-7
Molecular Formula C10H7ClO2S
Molecular Weight (g/mol) 226.674
MDL Number MFCD00052308
InChI Key QQKKTOPRRGBBCT-UHFFFAOYSA-N
Synonym 5-chlorobenzo b thiophene-3-acetic acid, 2-5-chlorobenzo b thiophen-3-yl acetic acid, 2-5-chloro-1-benzothiophen-3-yl acetic acid, benzo b thiophene-3-acetic acid, 5-chloro, 5-chloro-1-benzothien-3-yl acetic acid, 5-chloro-1-benzothiophen-3-yl acetic acid, maybridge4_002620, acmc-20am45
PubChem CID 205058
IUPAC Name 2-(5-chloro-1-benzothiophen-3-yl)acetic acid
SMILES C1=CC2=C(C=C1Cl)C(=CS2)CC(=O)O

Specifications

Melting Point 152°C to 155°C
Quantity 250 mg
Beilstein 1244795
Formula Weight 226.68
Percent Purity 97%
Chemical Name or Material 5-Chlorobenzo[b]thiophene-3-acetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.