missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chlorobenzofuroxan, 97%

Product Code. 11412407
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412407 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11412407 Supplier Alfa Aesar™ Supplier No. A14261.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 17348-69-5
Molecular Formula C6H3ClN2O2
Molecular Weight (g/mol) 170.552
MDL Number MFCD00068063
InChI Key DHPQXIQZZCNOLI-UHFFFAOYSA-N
Synonym 5-chlorobenzofuroxan, 5-chlorobenzofurazan 1-oxide, 2,1,3-benzoxadiazole, 5-chloro-, monooxide, 5-chloro-2,1,3-benzoxadiazol-1-ium-1-olate, 5-chloro-2,1,3-benzoxadiazole 1-oxide, acmc-20lz11, 5-chloro-beznofurazan oxide, 5-chlorobenzfurazan-1-oxide, 5-chlorobenzofurazan-1-oxide, 5-chlorobenzofurazane 1-oxide
PubChem CID 140222
IUPAC Name 5-chloro-1-oxido-2,1,3-benzoxadiazol-1-ium
SMILES C1=CC2=[N+](ON=C2C=C1Cl)[O-]

Specifications

Melting Point 45°C to 48°C
Quantity 25 g
Formula Weight 170.56
Percent Purity 97%
Chemical Name or Material 5-Chlorobenzofuroxan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.