missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorobenzotriazole, TRC

Product Code. 30352046
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30352046 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30352046 Supplier Toronto Research Chemicals Supplier No. TRCC382518500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chloro-1H-benzotriazole
CAS 94-97-3
Purity Grade Notes HPLC
Molecular Formula C6 H4 Cl N3
Quantity 500 mg
Synonym 1H-Benzotriazole, 5-chloro- (8CI,9CI), Benzotriazole, 5-chloro- (6CI,7CI), 6-Chloro-1H-benzotriazole, 5-Chloro-1H-benzo[d][1,2,3]triazole, 5-Chloro-1H-benzotriazole, 5-Chlorobenzotriazole, 6-Chlorobenzotriazole, NSC 16507, NSC 58361, 1H-Benzotriazole, 6-chloro-
InChI Formula InChI=1S/C6H4ClN3/c7-4-1-2-5-6(3-4)9-10-8-5/h1-3H,(H,8,9,10)
SMILES Clc1ccc2nn[nH]c2c1
IUPAC Name 6-chloro-1H-benzotriazole
Molecular Weight (g/mol) 153.56906
Formula Weight 153.0094
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.