missing translation for 'onlineSavingsMsg'
Learn More

5-Chloroindole-2-carboxylic Acid 98%, ACROS Organics™

Product Code. 10647252 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10647252 1 g Glass bottle 1g
10040910 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10647252 Supplier Thermo Scientific Acros Supplier No. 109320010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10517-21-2
Molecular Formula C9H5ClNO2
Molecular Weight (g/mol) 194.59
MDL Number MFCD00005613
InChI Key FUQOTYRCMBZFOL-UHFFFAOYSA-M
Synonym 5-chloroindole-2-carboxylic acid, 5-chloroindole-2-carboxylate, 1h-indole-2-carboxylic acid, 5-chloro, spectrum_000183, 5-chloroindoline-2-carboxylicacid, pubchem1670, acmc-1bztj, specplus_000670, spectrum2_000513, spectrum3_000783
PubChem CID 82693
IUPAC Name 5-chloro-1H-indole-2-carboxylate
SMILES [O-]C(=O)C1=CC2=CC(Cl)=CC=C2N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 283°C to 287°C
Color Beige to Pink or Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 195.6
Percent Purity ≥97.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material 5-Chloroindole-2-carboxylic acid, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.