missing translation for 'onlineSavingsMsg'
Learn More

5-(Chloromethyl)-2-[(tetrahydro-2H-pyran-4-yl)oxy]pyridine, TRC

Product Code. 30359754
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359754 25 mg -
30357877 5 mg 5mg
30367129 2.5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359754 Supplier Toronto Research Chemicals Supplier No. TRCC59840025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 906352-80-5
Molecular Formula C11H14NO2Cl
Molecular Weight (g/mol) 227.68
IUPAC Name 5-(chloromethyl)-2-(oxan-4-yloxy)pyridine
SMILES ClCc1ccc(OC2CCOCC2)nc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C11H14ClNO2/c12-7-9-1-2-11(13-8-9)15-10-3-5-14-6-4-10/h1-2,8,10H,3-7H2
Formula Weight 227.0713
Chemical Name or Material 5-(Chloromethyl)-2-[(tetrahydro-2H-pyran-4-yl)oxy]pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.