missing translation for 'onlineSavingsMsg'
Learn More

5-(Chloromethyl)-2-thien-2-ylpyrimidine, 97%, Thermo Scientific™

Product Code. 10491694
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10491694 1 g Amber Glass Bottle 1g
10401884 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10491694 Supplier Thermo Scientific Maybridge Supplier No. CC66524DA
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 926921-78-0
Molecular Formula C9H7ClN2S
Molecular Weight (g/mol) 210.68
MDL Number MFCD09879950
InChI Key HBGUDWOIHDHRCQ-UHFFFAOYSA-N
Synonym 5-chloromethyl-2-thien-2-ylpyrimidine, 5-chloromethyl-2-thiophen-2-yl pyrimidine, 2-5-chloromethyl pyrimidin-2-yl thiophene, 5-chloromethyl-2-thiophen-2-ylpyrimidine
PubChem CID 24229732
IUPAC Name 5-(chloromethyl)-2-thiophen-2-ylpyrimidine
SMILES ClCC1=CN=C(N=C1)C1=CC=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 210.69
Percent Purity 97%
Chemical Name or Material 5-(Chloromethyl)-2-thien-2-ylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.