missing translation for 'onlineSavingsMsg'
Learn More

5-Chloropent-1-yne, TRC

Product Code. 30358664
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358664 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30358664 Supplier Toronto Research Chemicals Supplier No. TRCC37509510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chloropent-1-yne
CAS 14267-92-6
Molecular Formula C5H7Cl
Quantity 10 g
Synonym (3-Chloropropyl)acetylene, 1-Chloro-4-pentyne, 4-Pentynyl Chloride, 5-Chloro-1-pentyne
InChI Formula InChI=1S/C5H7Cl/c1-2-3-4-5-6/h1H,3-5H2
SMILES ClCCCC#C
IUPAC Name 5-chloropent-1-yne
Molecular Weight (g/mol) 102.56
Formula Weight 102.0236
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.