missing translation for 'onlineSavingsMsg'
Learn More

5-Chloropyridine-2-boronic acid, 95%

Product Code. 15439257
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15439257 250 mg 250mg
15429257 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15439257 Supplier Thermo Scientific Alfa Aesar Supplier No. H52515.MD
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 652148-91-9 | C5H5BClNO2 | 157.36 g/mol

5-Chloropyridine-2-boronic acid was used as reagent in organic synthesis. Also used as phamaceutical Intermediates. Used in suzuki-Miyaura coupling processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloropyridine-2-boronic acid was used as reagent in organic synthesis. Also used as phamaceutical Intermediates. Used in suzuki-Miyaura coupling processes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652148-91-9
Molecular Formula C5H5BClNO2
Molecular Weight (g/mol) 157.36
MDL Number MFCD07368862
InChI Key JEGHCYRKSUGHJH-UHFFFAOYSA-N
Synonym 5-chloro-2-pyridineboronic acid, 5-chloropyridine-2-boronic acid, 5-chloropyridin-2-yl boronic acid, 2-borono-5-chloropyridine, 5-chloropyridine-2-boronicacid, 3-chloropyridine-6-boronic acid, 5-chloro-2-pyridyl boronic acid, boronic acid, 5-chloro-2-pyridinyl, pubchem13568, acmc-209npx
PubChem CID 22832086
IUPAC Name (5-chloropyridin-2-yl)boronic acid
SMILES B(C1=NC=C(C=C1)Cl)(O)O

Specifications

Quantity 250 mg
Formula Weight 157.37
Percent Purity 95%
Chemical Name or Material 5-Chloropyridine-2-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.