missing translation for 'onlineSavingsMsg'
Learn More

5-Chloropyridine-3-boronic acid, 95%, Thermo Scientific™

Product Code. 15403777
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15403777 250 mg 250mg
11322919 1 g 1g
11332919 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 15403777

Brand: Thermo Scientific Alfa Aesar H27756.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872041-85-5
Molecular Formula C5H5BClNO2
Molecular Weight (g/mol) 157.36
MDL Number MFCD06798243
InChI Key NJXYBTMCTZAUEE-UHFFFAOYSA-N
Synonym 5-chloropyridine-3-boronic acid, 5-chloropyridin-3-yl boronic acid, 5-chloro-3-pyridineboronic acid, 3-chloropyridine-5-boronic acid, 5-chloro-3-pyridinyl boronic acid, 5-chloro-3-pyridyl boronic acid, 5-chloropyridin-3-yl-3-boronic acid, 3-chloro-5-pyridyl boronic acid, 5-chloro-3-pyridineboronicacid, pubchem17397
PubChem CID 20111877
IUPAC Name (5-chloropyridin-3-yl)boronic acid
SMILES OB(O)C1=CC(Cl)=CN=C1

Specifications

Quantity 250 mg
Formula Weight 157.37
Percent Purity 95%
Chemical Name or Material 5-Chloropyridine-3-boronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.