missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorosaligenyl-N,N-diisopropylphosphoramidite, TRC

Product Code. 30349327
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30349327 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30349327 Supplier Toronto Research Chemicals Supplier No. TRCC3823335MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chlorosaligenyl-N,N-diisopropylphosphoramidite
CAS 1620086-77-2
Molecular Formula C13H19ClNO2P
Quantity 5 mg
Synonym 6-Chloro-N,N-bis(1-methylethyl)-4H-1,3,2-benzodioxaphosphorin-2-amine
InChI Formula InChI=1S/C13H19ClNO2P/c1-9(2)15(10(3)4)18-16-8-11-7-12(14)5-6-13(11)17-18/h5-7,9-10H,8H2,1-4H3
SMILES CC(C)N(C(C)C)P1OCc2cc(Cl)ccc2O1
IUPAC Name 6-chloro-N,N-di(propan-2-yl)-4H-1,3,2-benzodioxaphosphinin-2-amine
Molecular Weight (g/mol) 287.72
Formula Weight 287.0842
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.