missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorothiophene-2-boronic acid, 97+%

Product Code. 10031704 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10031704 5 g Glass bottle 5g
10017561 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10031704 Supplier Thermo Scientific Acros Supplier No. 359010050
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 162607-18-3 | C4H4BClO2S | 162.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162607-18-3
Molecular Formula C4H4BClO2S
Molecular Weight (g/mol) 162.39
MDL Number MFCD00236030
InChI Key JFCLNCVCDFUJPO-UHFFFAOYSA-N
Synonym 5-chlorothiophene-2-boronic acid, 5-chloro-2-thienylboronic acid, 5-chlorothiophen-2-yl boronic acid, 5-chloro-2-thiopheneboronic acid, 5-chlorothiophen-2-boronic acid, 5-chlorothiophen-2-yl boranediol, 5-chlorothiphene 2-boronic acid, 5-chloro-2-thienyl boronic acid, boronic acid, 5-chloro-2-thienyl, 5-chlorothiophene-2-boronicacid
PubChem CID 2734324
IUPAC Name (5-chlorothiophen-2-yl)boronic acid
SMILES OB(O)C1=CC=C(Cl)S1

Specifications

Melting Point 136.0°C to 140.0°C
Assay Percent Range 97+%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 162.4
Percent Purity 97+%
Chemical Name or Material 5-Chlorothiophene-2-boronic acid
compliance-icons
Product Identifier
  • 5-Chlorothiophene-2-boronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.