missing translation for 'onlineSavingsMsg'
Learn More

5-chlorothiophene-2-carboxylic acid, 98%, Thermo Scientific™

Product Code. 10002041 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10002041 5 g Glass bottle 5g
10026013 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10002041 Supplier Thermo Scientific Acros Supplier No. 352160050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 24065-33-6
Molecular Formula C5H3ClO2S
Molecular Weight (g/mol) 162.59
MDL Number MFCD00041426
InChI Key QZLSBOVWPHXCLT-UHFFFAOYSA-N
Synonym 5-chloro-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-chloro, 2-chloro-5-carboxythiophene, 5-chloro-2-thiophene carboxylic acid, pubchem7380, maybridge3_003717, acmc-209g8i, rivaroxaban related compound, 5-18-06-00177 beilstein handbook reference, ksc204m2j
PubChem CID 95048
IUPAC Name 5-chlorothiophene-2-carboxylic acid
SMILES OC(=O)C1=CC=C(Cl)S1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 146°C to 150°C
Color Cream-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 162.6
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-chlorothiophene-2-carboxylic acid
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • 5-Chlorothiophene-2-carboxylic acid
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.