missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorothiophene-2-sulfonyl chloride, 97%

Product Code. 11433774
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433774 5 g 5g
11423774 1 g 1g
11334659 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433774 Supplier Thermo Scientific Alfa Aesar Supplier No. A11348.06
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2766-74-7 | C4H2Cl2O2S2 | 217.08 g/mol

5-Chlorothiophene-2-sulfonyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs. It is used in the preparation of Notch-1-sparing γ-secretase inhibitors for the treatment of Alzheimer's disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Not miscible in water.

Notes
Moisture Sensitive. Store at 4°C. Incomaptible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2766-74-7
Molecular Formula C4H2Cl2O2S2
Molecular Weight (g/mol) 217.08
MDL Number MFCD00051667
InChI Key SORSTNOXGOXWAO-UHFFFAOYSA-N
Synonym 5-chlorothiophene-2-sulfonylchloride, 5-chloro-2-thiophenesulfonyl chloride, 5-chlorothiophene-2-sulphonyl chloride, 5-chlorothiophenesulphonyl chloride, 5-chloro-2-thienylsulfonyl chloride, 5-chloro-thiophene-2-sulfonyl chloride, 5-chlorothiophenesulfonyl chloride, 5-chlorothiophen-2-sulfonyl chloride, 5-chlorothiophene-2-sulphonylchloride, 2-thiophenesulfonyl chloride, 5-chloro
PubChem CID 2733925
IUPAC Name 5-chlorothiophene-2-sulfonyl chloride
SMILES ClC1=CC=C(S1)S(Cl)(=O)=O

Specifications

Density 1.623
Boiling Point 117°C to 120°C (14 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.586
Quantity 5 g
UN Number UN3265
Beilstein 130710
Sensitivity Moisture sensitive
Solubility Information Not miscible in water.
Formula Weight 217.09
Percent Purity 97%
Chemical Name or Material 5-Chlorothiophene-2-sulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.