missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorouracil, TRC

Product Code. 30138775
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138775 25 g 25g
30133364 10 g 10g
30004087 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138775 Supplier Toronto Research Chemicals Supplier No. TRCC42377525G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1820-81-1
Molecular Formula C4 H3 Cl N2 O2
Molecular Weight (g/mol) 146.53
Synonym 5-Chloropyrimidine-2,4(1H,3H)-dione, Fluorouracil Imp. E (EP), 2,4(1H,3H)-Pyrimidinedione, 5-chloro-, Uracil, 5-chloro- (6CI,7CI,8CI), 5-Chloro-2,4(1H,3H)-pyrimidinedione, 5-Chloro-2,4-dihydroxypyrimidine, 5-Chloro-2,4-pyrimidinedione, 5-Chloropyrimidine-2,4-diol, 5-Chlorouracil, NSC 28172
IUPAC Name 5-chloro-1H-pyrimidine-2,4-dione
SMILES ClC1=CNC(=O)NC1=O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C4H3ClN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)
Formula Weight 145.9883
Chemical Name or Material 5-Chlorouracil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.