missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorovaleroyl chloride, 98%

Product Code. 15422688
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15422688 50 g 50g
15432688 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15422688 Supplier Thermo Scientific Alfa Aesar Supplier No. H56237.18
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1575-61-7 | C5H8Cl2O | 155.02 g/mol

5-Chlorovaleroyl chloride (5-CVC) has been used in the development and validation of a GC-FID method for the analysis of low level impurities present in 5-CVC. It has been used in the synthesis of anion-exchange resins by acylation of polystyrene/divinylbenzene and subsequent amination with trimethylamine. It is commonly used as alkylating agent in the synthesis of pharmaceutical intermediates and other specialty chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chlorovaleroyl chloride (5-CVC) has been used in the development and validation of a GC-FID method for the analysis of low level impurities present in 5-CVC. It has been used in the synthesis of anion-exchange resins by acylation of polystyrene/divinylbenzene and subsequent amination with trimethylamine. It is commonly used as alkylating agent in the synthesis of pharmaceutical intermediates and other specialty chemicals.

Solubility
Reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, bases, alcohols, amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1575-61-7
Molecular Formula C5H8Cl2O
Molecular Weight (g/mol) 155.02
MDL Number MFCD00000758
InChI Key SVNNWKWHLOJLOK-UHFFFAOYSA-N
Synonym 5-chlorovaleryl chloride, valeryl chloride, 5-chloro, pentanoyl chloride, 5-chloro, 5-chlorovalerylchloride, 5-chlorovaleroyl chloride, pentanoylchloride, 5-chloro, chlorovaleryl chloride, pubchem10919, 5-chloro-pentanoylchloride, acmc-1cuj1
PubChem CID 74089
IUPAC Name 5-chloropentanoyl chloride
SMILES ClCCCCC(Cl)=O

Specifications

Density 1.2
Boiling Point 38°C to 39°C
Flash Point 91°C (196°F)
Linear Formula Cl(CH2)4COCl
Refractive Index 1.464
Quantity 50 g
UN Number UN3265
Beilstein 1745182
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 155.02
Percent Purity 98%
Chemical Name or Material 5-Chlorovaleroyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.