missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorovaleryl chloride, 96%

Product Code. 10429072 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
10mL
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10429072 10 mL Glass bottle 10mL
10061760 25 mL Glass bottle 25mL
10123110 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10429072 Supplier Thermo Scientific Acros Supplier No. 154770100
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1575-61-7 | C5H8Cl2O | 155.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1575-61-7
Molecular Formula C5H8Cl2O
Molecular Weight (g/mol) 155.02
MDL Number MFCD00000758
InChI Key SVNNWKWHLOJLOK-UHFFFAOYSA-N
Synonym 5-chlorovaleryl chloride, valeryl chloride, 5-chloro, pentanoyl chloride, 5-chloro, 5-chlorovalerylchloride, 5-chlorovaleroyl chloride, pentanoylchloride, 5-chloro, chlorovaleryl chloride, pubchem10919, 5-chloro-pentanoylchloride, acmc-1cuj1
PubChem CID 74089
IUPAC Name 5-chloropentanoyl chloride
SMILES ClCCCCC(Cl)=O

Spécification

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2000g/mL
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)4COCl
Refractive Index 1.4630 to 1.4650
Quantity 10 mL
Specific Gravity 1.2
Solubility Information Solubility in water: hydrolyses
Formula Weight 155.02
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 5-Chlorovaleryl chloride, 96%
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • 5-Chlorovaleryl chloride
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Mention de danger
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.