missing translation for 'onlineSavingsMsg'
Learn More

5α -Cholest-8(14)-en-3β-ol-15-one, 97%, Thermo Scientific™

Product Code. 10793897
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10793897 50 g 50g
10521893 1 g 1g
10069621 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10793897 Supplier Thermo Scientific Maybridge Supplier No. SB01709EE
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 50673-97-7
Molecular Formula C27H44O2
Molecular Weight (g/mol) 400.65
MDL Number MFCD09065035
InChI Key LINVVMHRTUSXHL-NDNSGUFDSA-N
Synonym colestolone, colestolonum, colestolona, 15-ketocholestene, unii-5p8396t5xf, 3beta-hydroxy-5alpha-cholest-8 14-en-15-one, 15-oxo-5alpha-cholest-8 14-en-3beta-ol, colestolone usan:inn, 5alpha-cholest-8 14-en-3-beta-ol-15-one, colestolonum inn-latin
PubChem CID 10046567
ChEBI CHEBI:61829
SMILES CC(C)CCCC(C)[C@H]1CC(=O)C2=C3CC[C@H]4C[C@@H](O)CC[C@]4(C)C3CC[C@]12C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 50 g
IUPAC Name (3S,5S,9R,10S,13R,17R)-3-hydroxy-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,3,4,5,6,7,9,11,12,16,17-dodecahydrocyclopenta[a]phenanthren-15-one
Formula Weight 400.64
Percent Purity 97%
Chemical Name or Material 5α -cholest-8(14)-en-3β-ol-15-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.