missing translation for 'onlineSavingsMsg'
Learn More

5α-Cholestan-3β-ol, MedChemExpress

Product Code. 16405126
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16405126 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16405126 Supplier Medchem Express Supplier No. HY107819
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 5α-Cholestan-3β-ol is a derivitized steroid compound.

5α-Cholestan-3β-ol is a derivitized steroid compound.

Specifications

Chemical Name or Material 5α-Cholestan-3β-ol
CAS 80-97-7
Color White
Purity Grade Notes Research
Molecular Formula C27H48O
Quantity 100 mg
Synonym Dihydrocholesterol 5α-Cholestanol NSC 18188
Solubility Information H2O : 1 mg/mL (2.57 mM; ultrasonic and warming and heat to 80°C) ∣DMSO : 1 mg/mL (2.57 mM; ultrasonic and warming and heat to 60°C)
SMILES C[C@@]12[C@](CC[C@]2([H])[C@H](C)CCCC(C)C)([H])[C@@]3([H])[C@]([C@@]4([C@@](C[C@@H](O)CC4)([H])CC3)C)([H])CC1
Molecular Weight (g/mol) 388.67
Formula Weight 388.67
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.