missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ 5-CR 6G, SE (5-Carboxyrhodamine 6G, Succinimidyl Ester), single isomer
Description
The amine-reactive form of 5-CR 6G is used for oligonucleotide labeling and automated DNA sequencing applications. Conjugates of this dye have longer-wavelength spectra than the spectra of Lisaamine™ rhodamine B conjugates, but somewhat shorter-wavelength spectra than those of Texas Red™ conjugates.
Specifications
Specifications
| Quantity | 5 mg |
| Product Type | 5-CR 6G SE |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Amine |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Active Ester, Succinimidyl Ester |
| Label or Dye | Rhodamine 6G |
| Label Type | Classic Dyes |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?