missing translation for 'onlineSavingsMsg'
Learn More

5-Desamino 5-Oxo-2,5-dihydro Lamotrigine, TRC

Product Code. 18051430
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18051430 25 mg 25mg
18050930 10 mg 10mg
18030060 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18051430 Supplier Toronto Research Chemicals Supplier No. TRCD28847025MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 252186-78-0
Molecular Formula C9 H6 Cl2 N4 O
Molecular Weight (g/mol) 257.08
Synonym 3-Amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-5(4H)-one, 1,2,4-Triazin-5(2H)-one, 3-amino-6-(2,3-dichlorophenyl)-, 3-Amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-5(2H)-one, 5-Desamino 5-Oxo-2,5-dihydro Lamotrigine
IUPAC Name 3-amino-6-(2,3-dichlorophenyl)-4H-1,2,4-triazin-5-one
SMILES NC1=NN=C(C(=O)N1)c2cccc(Cl)c2Cl

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C9H6Cl2N4O/c10-5-3-1-2-4(6(5)11)7-8(16)13-9(12)15-14-7/h1-3H,(H3,12,13,15,16)
Formula Weight 255.9919
Percent Purity >95%
Chemical Name or Material 3-Amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-5(4H)-one
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.