missing translation for 'onlineSavingsMsg'
Learn More

5-(Di-tert-butylphosphino)-1',3',5'-triphenyl-1'H-1,4-bipyrazole, 97%

Product Code. 11985951 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11985951 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11985951 Supplier Thermo Scientific Acros Supplier No. 435160010
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 894086-00-1 | C32H35N4P | 506.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 894086-00-1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 169.0°C to 173.0°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C32H35N4P
Quantity 1 g
Synonym bippyphos, t-bu-bippyphos, 5-di-tert-butylphosphino-1',3',5'-triphenyl-1'h-1,4'-bipyrazole, 5-di-tert-butylphosphino-1', 3', 5'-triphenyl-1'h-1,4' bipyrazole, 5-di-tert-butylphosphino-1',3',5'-triphenyl-1'h-1,4' bipyrazole, 5-di-t-butylphosphino-1',3',5'-triphenyl-1,4'-bi-1h-pyrazole, 5-bis tert-butyl phosphino-1',3',5'-triphenyl-1,4'-bi-1h-pyrazole, 5-di-tert-butylphosphanyl-1',3',5'-triphenyl-1,4'-bipyrazole, 1',3',5'-triphenyl-5-ditert-butylphosphino-1,4'-bi 1h-pyrazole, 5-di-tert-butyl-phosphanyl-1',3',5'-triphenyl-1'h-1,4' bipyrazolyl
InChI Key PTXJGGGNGMPMBG-UHFFFAOYSA-N
SMILES CC(C)(C)P(C1=CC=NN1C2=C(N(N=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5)C(C)(C)C
IUPAC Name ditert-butyl-[2-(1,3,5-triphenylpyrazol-4-yl)pyrazol-3-yl]phosphane
Molecular Weight (g/mol) 506.62
PubChem CID 11547931
Formula Weight 506.62
Percent Purity 97%
Physical Form Crystalline Powder or Solid
Chemical Name or Material 5-(Di-tert-Butylphosphino)-1',3',5'-triphenyl-1'H-1,4-bipyrazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.