missing translation for 'onlineSavingsMsg'
Learn More

5-(Dimethylamino)naphthalene-1-sulfonyl chloride, 97%, BLD Pharm™

Product Code. 31104597
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31104597 25 g
31093245 250 mg
31092429 5 g
31104091 10 g
31097981 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31104597 Supplier BLD Pharm Supplier No. BD1385125g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 605-65-2
Molecular Formula C12H12ClNO2S
MDL Number MFCD00003985
Quantity 25 g
SMILES O=S(C1=C2C=CC=C(N(C)C)C2=CC=C1)(Cl)=O
Molecular Weight (g/mol) 269.75
Percent Purity 97%
Chemical Name or Material 5-(Dimethylamino)naphthalene-1-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.