missing translation for 'onlineSavingsMsg'
Learn More

5-Ethyl-2-furaldehyde, 98%, Thermo Scientific™

Product Code. 10778742 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10778742 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10778742 Supplier Thermo Scientific Acros Supplier No. 336550050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 23074-10-4
CAS Min % 97.5
CAS Max % 100.0
Density 1.0500g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H8O2
Refractive Index 1.5215 to 1.5235 (20°C, 589nm)
MDL Number MFCD00030605
Quantity 5g
Synonym 5-ethyl-2-furaldehyde, 5-ethylfurfural, 2-furancarboxaldehyde, 5-ethyl, 5-ethyl-furan-2-carbaldehyde, 2-furancarboxaldehyde,5-ethyl, 5-ethylfurfuraldehyde, acmc-1cc1a, 5-ethyl-2-furancarboxaldehyde, 5-ethylfuran-2-carboxaldehyde
InChI Key XADGZBXFWQHBDB-UHFFFAOYSA-N
SMILES CCC1=CC=C(O1)C=O
IUPAC Name 5-ethylfuran-2-carbaldehyde
Molecular Weight (g/mol) 124.14
PubChem CID 89989
Formula Weight 124.14
Percent Purity ≥97.5%
Chemical Name or Material 5-Ethyl-2-furaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.