missing translation for 'onlineSavingsMsg'
Learn More

5-Ethyl-4-hydroxy-2-methyl-3(2H)-furanone (mixture of isomers) ( >80%), TRC

Product Code. 30381729
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381729 1 g -
30373365 10 g 10g
30385823 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381729 Supplier Toronto Research Chemicals Supplier No. TRCE9455981G
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27538-09-6
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.15
Synonym 3(2H)-Furanone, 5-ethyl-4-hydroxy-2-methyl-, 5-Ethyl-4-hydroxy-2-methyl-3(2H)-furanone, 2-Ethyl-5-methyl-4,5-dihydrofuran-3-ol-4-one, 2-Methyl-4-hydroxy-5-ethylfuran-3-one, 2-Methyl-5-ethyl-4-hydroxy-2,3-dihydrofuran-3-one, 4-Hydroxy-2-methyl-5-ethyl-2,3-dihydrofuran-3-one, 4-Hydroxy-5-ethyl-2-methyl-3(2H)-furanone, 5-Ethyl-2,3-dihydro-4-hydroxy-2-methyl-3-furanone, 5-Ethyl-2-methyl-4-hydroxy-2,3-dihydrofuran-3-one, HEMF, Homofuraneol, Homofuronol
IUPAC Name 5-ethyl-4-hydroxy-2-methylfuran-3-one
SMILES CCC1=C(O)C(=O)C(C)O1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H10O3/c1-3-5-7(9)6(8)4(2)10-5/h4,9H,3H2,1-2H3
Formula Weight 142.063
Chemical Name or Material 5-Ethyl-4-hydroxy-2-methyl-3(2H)-furanone (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.