missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methoxybenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15465507
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465507 5 g 5g
15475507 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465507 Supplier Thermo Scientific Alfa Aesar Supplier No. H32748.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 179897-94-0
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.95
MDL Number MFCD01863526
InChI Key CCQKIRUMTHHPSX-UHFFFAOYSA-N
Synonym 5-fluoro-2-methoxyphenyl boronic acid, 2-methoxy-5-fluorophenylboronic acid, 5-fluoro-2-methoxybenzeneboronic acid, 2-methoxy-5-fluorobenzeneboronic acid, 5-fluoro-2-methoxyphenylboronicacid, 2-borono-4-fluoroanisole, 3-fluoro-6-methoxybenzeneboronic acid, 5-fluoro-2-methoxyphenyl boranediol, 4-fluoroanisole-2-boronic acid, boronic acid, 5-fluoro-2-methoxyphenyl
PubChem CID 2782673
IUPAC Name (5-fluoro-2-methoxyphenyl)boronic acid
SMILES COC1=CC=C(F)C=C1B(O)O

Specifica

Quantity 5 g
Formula Weight 169.95
Percent Purity 98%
Chemical Name or Material 5-Fluoro-2-methoxybenzeneboronic acid
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.