missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-nitroaniline, 97%

Product Code. 15408388
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
15408388 5 g 5g
15418388 25 g 25g
15428388 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 15408388

Brand: Thermo Scientific Alfa Aesar H61806.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2369-11-1 | C6H5FN2O2 | 156.12 g/mol

5-Fluoro-2-nitroaniline as a intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoro-2-nitroaniline as a intermediate in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2369-11-1
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.12
MDL Number MFCD00034065
InChI Key PEDMFCHWOVJDNW-UHFFFAOYSA-N
IUPAC Name 5-fluoro-2-nitroaniline

Spezifikation

Quantity 5 g
UN Number UN2811
Synonym 4-bromo-2,6-xylenol, phenol, 4-bromo-2,6-dimethyl, 2,6-dimethyl-4-bromophenol, 2,6-xylenol, 4-bromo, pubchem9579, 2, 4-bromo, acmc-2097aq, 4-bromo-2,6 dimethylphenol, 5-bromo-2-hydroxy-m-xylene, ksc495g0d
Solubility Information Slightly soluble in water.
SMILES NC1=CC(F)=CC=C1[N+]([O-])=O
PubChem CID 16919
Formula Weight 156.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Fluoro-2-nitroaniline

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.