missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride, 97%, Thermo Scientific™

Product Code. 10453162
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10453162 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10453162 Supplier Thermo Scientific Maybridge Supplier No. MO07029DA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 84163-16-6
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C12H14ClFN2O
MDL Number MFCD06200879
Quantity 1 g
Synonym 5-fluoro-3-piperidin-4-yl benzo d isoxazole hydrochloride, 5-fluoro-3-piperidin-4-yl-1,2-benzisoxazole hydrochloride, 5-fluoro-3-4-piperidinyl-1,2-benzisoxazole hydrochloride, 5-fluoro-3-piperidin-4-yl-1,2-benzoxazole hydrochloride, 5-fluoro-3-piperidin-4-yl benzo d isoxazole hcl, 4-5-fluoro-1,2-benzisoxazol-3-yl piperidine hydrochloride, 1,2-benzisoxazole,5-fluoro-3-4-piperidinyl-, hydrochloride, pubchem11469, 5-fluoro-3-piperidin-4-yl benzisoxazole hcl salt, 5-fluoro-3-4-piperidyl benzo d isoxazole, chloride
InChI Key HSOJFILHZZEWJT-UHFFFAOYSA-N
SMILES C1CNCCC1C2=NOC3=C2C=C(C=C3)F.Cl
IUPAC Name 5-fluoro-3-piperidin-4-yl-1,2-benzoxazole;hydrochloride
Molecular Weight (g/mol) 256.705
PubChem CID 2794754
Formula Weight 256.71
Percent Purity 97%
Chemical Name or Material 5-Fluoro-3-(4-piperidinyl)-1, 2-benzisoxazole hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.