missing translation for 'onlineSavingsMsg'
Learn More

Abcam 5-Fluoroorotic Acid Hydrate, >99% Purity, Solid, 1 g

Product Code. 32119397 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32119397 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32119397 Supplier Abcam Supplier No. ab1462451g
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity selective agent for yeast molecular genetics.

  • Purity: >99%
  • Molecular Weight: 174.1 g/mol
  • Storage Temperature: Store at -20°C
  • Shipping Temperature: Ambient - Can Ship with Ice
  • Form: Solid
  • Applications: Selective agent in yeast molecular genetics, useful in the selection of orotidine-5'-phosphate decarboxylase mutants of Saccharomyces cerevisiae
  • Composition: 100% 5-fluoro-2,4-dioxo-1H-pyrimidine-6-carboxylic acid; hydrate

The product is subject to the following: Abcam Restricted Use Statement

Spezifikation

Buffer 100% 5-fluoro-2,4-dioxo-1H-pyrimidine-6-carboxylic acid, hydrate
Content And Storage Store at -20°C.
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule Orotidine-5¹-phosphate Decarboxylase
Formulation 100% 5-fluoro-2,4-dioxo-1H-pyrimidine-6-carboxylic acid;hydrate
Quantity 1 g
For Use With (Application) Yeast molecular genetics, selection of orotidine-5'-phosphate decarboxylase mutants
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.