missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoropyridine-3-boronic acid, 98%

Product Code. 15489627
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15489627 250 mg 250mg
15479627 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15489627 Supplier Thermo Scientific Alfa Aesar Supplier No. H52860.MD
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 872041-86-6 | C5H5BFNO2 | 140.908 g/mol

5-Fluoropyridine-3-boronic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoropyridine-3-boronic acid is used as pharmaceutical intermediate.

Solubility
Soluble in hot methanol.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872041-86-6
Molecular Formula C5H5BFNO2
Molecular Weight (g/mol) 140.908
MDL Number MFCD07368243
InChI Key FVEDGBRHTGXPOK-UHFFFAOYSA-N
Synonym 5-fluoropyridine-3-boronic acid, 5-fluoropyridin-3-yl boronic acid, 5-fluoropyridin-3-yl-3-boronic acid, 5-fluoropyridine-3-boronicacid, 5-fluoro-3-pyridylboronic acid, 3-fluoropyridine-5-boronic acid, 5-fluoro-3-pyridyl boronic acid, acmc-209qir, boric acid fluoride-3-5, 3-borono-5-fluoropyridine
PubChem CID 44717403
IUPAC Name (5-fluoropyridin-3-yl)boronic acid
SMILES B(C1=CC(=CN=C1)F)(O)O

Specifications

Melting Point 233°C to 236°C
Quantity 250 mg
Solubility Information Soluble in hot methanol.
Formula Weight 140.91
Percent Purity 98%
Chemical Name or Material 5-Fluoropyridine-3-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.