missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroquinazolin-4-ol, 97%, Thermo Scientific™

Product Code. 10671416
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10671416 5 g 5g
10019091 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10671416 Supplier Thermo Scientific Maybridge Supplier No. BTB04061DE
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 436-72-6
Molecular Formula C8H5FN2O
Molecular Weight (g/mol) 164.14
MDL Number MFCD00173674
InChI Key UXEZULVIMJVIFB-UHFFFAOYSA-N
Synonym 5-fluoro-4-hydroxyquinazoline, 5-fluoroquinazolin-4-ol, 5-fluoroquinazolin-4 1h-one, 5-fluoroquinazolin-4 3h-one, 5-fluoro-4-quinazolone, 5-fluoro-3h-quinazolin-4-one, 4 1h-quinazolinone, 5-fluoro, pubchem6307, 5-fluoroquinazolin-4-one, acmc-209jv3
PubChem CID 2737331
SMILES FC1=C2C(NC=NC2=O)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
IUPAC Name 5-fluoro-1H-quinazolin-4-one
Formula Weight 164.14
Percent Purity 97%
Chemical Name or Material 5-Fluoroquinazolin-4-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.