missing translation for 'onlineSavingsMsg'
Learn More

5-Formyl-2-methylpyrimidine, TRC

Product Code. 30397227
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397227 1 g 1g
30403733 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30397227 Supplier Toronto Research Chemicals Supplier No. TRCF7008101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Formyl-2-methylpyrimidine
CAS 90905-33-2
Molecular Formula C6 H6 N2 O
Quantity 1 g
Synonym 5-Pyrimidinecarboxaldehyde, 2-methyl-, 2-Methyl-5-pyrimidinecarboxaldehyde, 5-Formyl-2-methylpyrimidine, 2-Methyl-5-pyrimidinecarbaldehyde, 2-Methylpyrimidine-5-carboxaldehyde
InChI Formula InChI=1S/C6H6N2O/c1-5-7-2-6(4-9)3-8-5/h2-4H,1H3
SMILES Cc1ncc(C=O)cn1
IUPAC Name 2-methylpyrimidine-5-carbaldehyde
Molecular Weight (g/mol) 122.12
Formula Weight 122.048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.