missing translation for 'onlineSavingsMsg'
Learn More

5-Formylthiophene-2-boronic acid, 97%

Product Code. 11422300
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422300 1 g 1g
11432300 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11422300 Supplier Thermo Scientific Alfa Aesar Supplier No. H29289.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4347-33-5 | C5H5BO3S | 155.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4347-33-5
Molecular Formula C5H5BO3S
Molecular Weight (g/mol) 155.96
MDL Number MFCD02093666
InChI Key DEQOVKFWRPOPQP-UHFFFAOYSA-N
Synonym 5-formyl-2-thiopheneboronic acid, 5-formylthiophen-2-boronic acid, 5-formylthiophen-2-yl boronic acid, 5-formylthiophene-2-boronic acid, 2-formylthiophene-5-boronic acid, 5-formylthienyl-2-boronic acid, 5-formyl-2-thienyl boronic acid, boronic acid, 5-formyl-2-thienyl, 5-formyl-2-thiopheneboronicacid, pubchem1722
PubChem CID 2773430
IUPAC Name (5-formylthiophen-2-yl)boronic acid
SMILES OB(O)C1=CC=C(S1)C=O

Specifications

Melting Point ∽124°C (decomposition)
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 155.97
Percent Purity 97%
Chemical Name or Material 5-Formylthiophene-2-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.