missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 5'-Guanylic acid disodium salt

Product Code. 16479346
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479346 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479346 Supplier MedChemExpress Supplier No. HYW010970
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 5'-Guanylic acid disodium salt (5'-GMP disodium salt) is composed of guanine, ribose, and phosphate moieties and it is a nucleotide monomer in messenger RNA. Guanosine derivatives are involved in intracellular signal transduction and have been identified in repetitive genomic sequences in telomeres, in ribosomal DNA, immunoglobulin heavy‐chain switch regions, and in the control regions of proto-oncogenes.

Specifications

Molecular Weight (g/mol) 407.18 g/mol
Product Type Natural Products
Content And Storage 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Shipping Condition Room temperature
Purity 97.0%
Quantity 100 mg
Solubility Information H2O : 100 mg/mL (245.59 mM∣Need ultrasonic) ∣DMSO : <1 mg/mL (ultrasonic∣warming∣heat to 80°C) (insoluble or slightly soluble)
For Use With (Application) Metabolism-protein/nucleotide metabolism
CAS 5550-12-9
Synonym 5'-GMP disodium salt∣5'-guanosine monophosphate disodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.