missing translation for 'onlineSavingsMsg'
Learn More

5-Hexen-2-one, 98%

Product Code. p-7026488
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11420697

Brand: Thermo Scientific Alfa Aesar B22505.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-49-9 | C6H10O | 98.145 g/mol

Allyl Acetone is used as a nature identical flavor and fragrance. And it is also used to produce 2-methyl-hex-5-en-2-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl Acetone is used as a nature identical flavor and fragrance. And it is also used to produce 2-methyl-hex-5-en-2-ol.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-49-9
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00008793
InChI Key RNDVGJZUHCKENF-UHFFFAOYSA-N
Synonym 5-hexen-2-one, allylacetone, 5-hexene-2-one, 1-hexen-5-one, 3-buten-1-yl methyl ketone, unii-xwm6rl9yei, xwm6rl9yei, 1-hexene-5-one, 5-oxo-1-hexene, allyl acetone
PubChem CID 7989
IUPAC Name hex-5-en-2-one
SMILES CC(=O)CCC=C

Specifications

Density 0.847
Boiling Point 127°C to 128°C
Flash Point 23°C (73°F)
Odor Like Ketone
Refractive Index 1.42
Quantity 5 g
UN Number UN1224
Beilstein 1699137
Solubility Information Soluble in water.
Formula Weight 98.15
Percent Purity ≥98%
Chemical Name or Material 5-Hexen-2-one
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.