missing translation for 'onlineSavingsMsg'
Learn More

5-Hexen-2-one, 98%

Product Code. 10764241 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
20 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
20mL
5mL
This item is not returnable. View return policy

Product Code. 10764241

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-49-9 | C6H10O | 98.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-49-9
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.14
MDL Number MFCD00008793
InChI Key RNDVGJZUHCKENF-UHFFFAOYSA-N
Synonym 5-hexen-2-one, allylacetone, 5-hexene-2-one, 1-hexen-5-one, 3-buten-1-yl methyl ketone, unii-xwm6rl9yei, xwm6rl9yei, 1-hexene-5-one, 5-oxo-1-hexene, allyl acetone
PubChem CID 7989
IUPAC Name hex-5-en-2-one
SMILES CC(=O)CCC=C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 0.8400g/mL
Boiling Point 128°C to 129°C
Flash Point 23°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2CH2COCH3
Refractive Index 1.4190 to 1.421
Quantity 20 mL
Beilstein 01, 734
Specific Gravity 0.84
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in organic solvents
Formula Weight 98.14
Percent Purity 98%
Chemical Name or Material 5-Hexen-2-one
Show More Show Less
compliance-icons
Product Identifier
  • 5-Hexen-2-one
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.