missing translation for 'onlineSavingsMsg'
Learn More

5-Hexenenitrile, 97%, Thermo Scientific™

Product Code. 11343106
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11343106

Brand: Thermo Scientific Alfa Aesar L10968.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Hexenenitrile, is used as a natural substances and extractives.

Solubility
Insoluble in water. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5048-19-1
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.145
MDL Number MFCD00067056
InChI Key UNAQSRLBVVDYGP-UHFFFAOYSA-N
Synonym 5-hexenenitrile, 5-cyano-1-pentene, 4-pentenyl cyanide, hex-1-ene-6-nitrile, acmc-209kmu
PubChem CID 123222
IUPAC Name hex-5-enenitrile
SMILES C=CCCCC#N

Specifications

Boiling Point 160°C to 162°C
Flash Point 40°C (104°F)
Odor Recognizable
Quantity 1 g
UN Number UN1992
Beilstein 1739773
Formula Weight 95.15
Percent Purity 97%
Chemical Name or Material 5-Hexenenitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.