missing translation for 'onlineSavingsMsg'
Learn More

5-Hexynenitrile, 98%

Product Code. 10781925 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10781925 5 g Glass bottle 5g
10135800 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10781925 Supplier Thermo Scientific Supplier No. 307370050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14918-21-9 | C6H7N | 93.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 14918-21-9
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
MDL Number MFCD00001978
InChI Key JZYKFLLRVPPISG-UHFFFAOYSA-N
Synonym 5-hexynenitrile, 5-cyano-1-pentyne, acmc-1c81n, 5-hexynenitrile 1g
PubChem CID 139852
IUPAC Name hex-5-ynenitrile
SMILES C#CCCCC#N

Specifikationer

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Amber
Density 0.8800g/mL
Boiling Point 115°C to 117°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HC∽C(CH2)3CN
Refractive Index 1.4390 to 1.441
Quantity 5 g
Specific Gravity 0.88
Formula Weight 93.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Hexynenitrile
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.